Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RGS10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | RGS10 |
---|---|
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15539620
![]() |
Novus Biologicals
NBP15539620UL |
20 μL |
Each for $206.00
|
|
|||||
NBP155396
![]() |
Novus Biologicals
NBP155396 |
100 μL |
Each for $487.50
|
|
|||||
Description
RGS10 Polyclonal specifically detects RGS10 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
RGS10 | |
Polyclonal | |
Rabbit | |
Q96GN0 | |
6001 | |
Synthetic peptides corresponding to RGS10 (regulator of G-protein signalling 10) The peptide sequence was selected from the middle region of RGS10. Peptide sequence DQIFNLMKYDSYSRFLKSDLFLKHKRTEEEEEDLPDAQTAAKRASRIYNT. | |
Primary |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
regulator of G-protein signaling 10, regulator of G-protein signalling 10 | |
RGS10 | |
IgG | |
21 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title