Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS11 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056658 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056658 0.2 mL
NB10056658T 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056658 Supplier Novus Biologicals Supplier No. NB10056658
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

RGS11 Polyclonal specifically detects RGS11 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen RGS11
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:200-1:1000
Formulation PBS containing 0.2% gelatin with 0.05% Sodium Azide
Gene Alias regulator of G-protein signaling 11, regulator of G-protein signalling 11, RS11
Gene Symbols RGS11
Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 201-217 (SADFHKREIEYFRKALG) of human RGS11.
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8786
Test Specificity An approx. 48 kDa band is observed.
Target Species Human, Mouse, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.