Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RGS13 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$428.79 - $684.12
Specifications
| Antigen | RGS13 |
|---|---|
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
RGS13 Polyclonal antibody specifically detects RGS13 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| RGS13 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS, pH 7.2, 40% glycerol | |
| 6003 | |
| IgG | |
| Affinity purified |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| MGC17173, regulator of G-protein signaling 13, regulator of G-protein signalling 13 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: TYKKIASRWSRISRAKKLYKIYIQPQSPREINIDSSTRETIIRNIQEPTETCFEEAQK | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title