Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS14 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220147 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220147 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220147 Supplier Novus Biologicals Supplier No. NBP220147
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RGS14 Polyclonal specifically detects RGS14 in Human samples. It is validated for Western Blot.

Specifications

Antigen RGS14
Applications Western Blot
Classification Polyclonal
Concentration 0.79mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. O43566
Gene Alias regulator of G-protein signaling 14, regulator of G-protein signalling 14
Gene Symbols RGS14
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RGS14. The exact sequence is proprietary.
Molecular Weight of Antigen 61 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10636
Test Specificity Human. Species reactivity based on sequence identity: Bovine (84%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.