Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS18 Antibody - BSA Free
SDP

Catalog No. NB300335 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300335 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300335 Supplier Novus Biologicals Supplier No. NB300335

Chicken Polyclonal Antibody

RGS18 Polyclonal specifically detects RGS18 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoaffinity Purification.

Specifications

Antigen RGS18
Applications Western Blot, Affinity Purification
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:200 - 1:1500, Immunoaffinity Purification
Formulation PBS (pH 7.2) with 0.02% Sodium Azide
Gene Alias regulator of G-protein signaling 18, regulator of G-protein signalling 13, RGS13regulator of G-protein signalling 18
Gene Symbols RGS18
Host Species Chicken
Immunogen Synthetic peptide: INMCESKEKT FFKLIHGSGK EETSKEAKIR AKEKRNRLSL LVQKPEFHED TRSSRSGHLA KETRVSPEEA VKWGESFDKL LSHRDGLEAF, corresponding to amino acids 11-100 of RGS18.
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 64407
Test Specificity RGS18
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.