Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS18 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224417SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224417SS 0.025 mL
NBP224417 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224417SS Supplier Novus Biologicals Supplier No. NBP224417SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

RGS18 Polyclonal specifically detects RGS18 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen RGS18
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:250-1: 1000, Immunocytochemistry/Immunofluorescence 1: 100
Formulation 0.1 ml crude whole Rabbit sera with 50% Glycerol with 0.025% Sodium Azide
Gene Accession No. Q9NS28
Gene Alias regulator of G-protein signaling 18, regulator of G-protein signalling 13, RGS13regulator of G-protein signalling 18
Gene Symbols RGS18
Host Species Rabbit
Immunogen Within the range of amino acids 30-62 of human RGS18 protein were used as the immunogen.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 64407
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.