Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS19 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP224516SS

 View more versions of this product

Catalog No. NBP224516SS


Only null left
Add to Cart

Description

Description

RGS19 Polyclonal specifically detects RGS19 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

RGS19
Polyclonal
Unconjugated
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
G alpha interacting protein, GAIPG protein signalling regulator 19, G-alpha-interacting protein, GNAI3IP, guanine nucleotide binding protein alpha inhibiting activity polypeptide 3interacting protein, regulator of G-protein signaling 19, regulator of G-protein signalling 19, RGSGAIP
Rabbit
Affinity Purified
RUO
Primary
The amino acid sequence used as immunogen is 100% homologous in human, 95% homologous in mouse and rat and 94% homologous in monkey.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot, Immunohistochemistry (Paraffin)
0.5 mg/mL
Western Blot 0.5-2 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
P49795
RGS19
A portion of amino acids 1-50 of human RGS19 was used as the immunogen.
0.025 mg
Autophagy, Signal Transduction
10287
Human, Mouse
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only