Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RGS19 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | RGS19 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.5-2 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224516SS
![]() |
Novus Biologicals
NBP224516SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP224516
![]() |
Novus Biologicals
NBP224516 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
RGS19 Polyclonal specifically detects RGS19 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RGS19 | |
Western Blot 0.5-2 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
Polyclonal | |
Rabbit | |
Autophagy, Signal Transduction | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
G alpha interacting protein, GAIPG protein signalling regulator 19, G-alpha-interacting protein, GNAI3IP, guanine nucleotide binding protein alpha inhibiting activity polypeptide 3interacting protein, regulator of G-protein signaling 19, regulator of G-protein signalling 19, RGSGAIP | |
RGS19 | |
IgG | |
Affinity Purified | |
The amino acid sequence used as immunogen is 100% homologous in human, 95% homologous in mouse and rat and 94% homologous in monkey. |
0.5 mg/mL | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse | |
P49795 | |
10287 | |
A portion of amino acids 1-50 of human RGS19 was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title