Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS20 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen RGS20
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP154885
SDP
View Documents
Novus Biologicals
NBP154885
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

RGS20 Polyclonal specifically detects RGS20 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

RGS20
Polyclonal
Purified
RUO
O76081
8601
Synthetic peptide directed towards the middle region of human RGS20 (NP_003693). Peptide sequence NAFREFLRTEFSEENMLFWMACEELKKEANKNIIEEKARIIYEDYISILS.
Primary
25 kDa
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Signal Transduction
G(z)GAP, Gz-GAP, Gz-selective GTPase-activating protein, regulator of G-protein signaling 20, Regulator of G-protein signaling Z1, regulator of G-protein signalling 20, Regulator of Gz-selective protein signaling 1, RGSZ1g(z)GAP, ZGAP1gz-GAP
RGS20
IgG
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.