Learn More
Description
Specifications
Specifications
| Antigen | RGS20 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS, 2% Sucrose with 0.09% Sodium Azide |
| Gene Accession No. | O76081 |
| Gene Alias | G(z)GAP, Gz-GAP, Gz-selective GTPase-activating protein, regulator of G-protein signaling 20, Regulator of G-protein signaling Z1, regulator of G-protein signalling 20, Regulator of Gz-selective protein signaling 1, RGSZ1g(z)GAP, ZGAP1gz-GAP |
| Gene Symbols | RGS20 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
