Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS22 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB165201 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165201 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB165201 Supplier Novus Biologicals Supplier No. NBP281776UV
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RGS22 Polyclonal antibody specifically detects RGS22 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen RGS22
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias DKFZp434I092, FLJ40080, FLJ75004, MGC102908, PRTD-NY2, regulator of G-protein signaling 22, regulator of G-protein signalling 22
Host Species Rabbit
Immunogen RGS22 antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human RGS22. The immunogen is located within amino acids 870 - 920 of RGS22. Amino Acid Squence: TDIEQFRRITYRDRNQR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 26166
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.