Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RGS5 Antibody (CL5568), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261419
Description
RGS5 Monoclonal specifically detects RGS5 in Human samples. It is validated for Western Blot.Specifications
RGS5 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
RGS5 | |
This antibody was developed against a synthetic peptide corresponding to amino acids: GDLVIPYNEKPEKPAKTQK | |
100 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry (Paraffin) | |
CL5568 | |
Western Blot 1:500 - 1:1000, Immunohistochemistry-Paraffin | |
MST092, MST106, MST129, MSTP032, MSTP092, MSTP106, MSTP129, regulator of G-protein signaling 5, regulator of G-protein signalling 5 | |
Mouse | |
Protein A purified | |
Signal Transduction | |
8490 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction