Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RGS5 Antibody (CL5568), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $610.00
Specifications
Antigen | RGS5 |
---|---|
Clone | CL5568 |
Dilution | Western Blot 1:500 - 1:1000, Immunohistochemistry-Paraffin |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
RGS5 Monoclonal specifically detects RGS5 in Human samples. It is validated for Western Blot.Specifications
RGS5 | |
Western Blot 1:500 - 1:1000, Immunohistochemistry-Paraffin | |
Unconjugated | |
Mouse | |
MST092, MST106, MST129, MSTP032, MSTP092, MSTP106, MSTP129, regulator of G-protein signaling 5, regulator of G-protein signalling 5 | |
RGS5 | |
IgG1 | |
Protein A purified |
CL5568 | |
Monoclonal | |
Purified | |
Signal Transduction | |
8490 | |
This antibody was developed against a synthetic peptide corresponding to amino acids: GDLVIPYNEKPEKPAKTQK | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title