Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS5 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224945 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224945 0.1 mL
NBP224945SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224945 Supplier Novus Biologicals Supplier No. NBP224945
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RGS5 Polyclonal specifically detects RGS5 in Human, Rat, Porcine, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen RGS5
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:250-1: 1000, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200
Formulation 0.1 ml crude whole Rabbit sera with 50% Glycerol with 0.025% Sodium Azide
Gene Accession No. O15539
Gene Alias MST092, MST106, MST129, MSTP032, MSTP092, MSTP106, MSTP129, regulator of G-protein signaling 5, regulator of G-protein signalling 5
Gene Symbols RGS5
Host Species Rabbit
Immunogen Within the range of amino acids 169-181 of human RGS5 were used as immunogen.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8490
Test Specificity This antibody is reactive to Chimpanzee, Dog, Human, Monkey, Mouse, Pig, Rat.
Target Species Human, Rat, Pig, Canine, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.