Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS5 Antibody (OTI1C1), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N27387APCY7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N27387APCY7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N27387APCY7 Supplier Novus Biologicals Supplier No. NBP273887APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RGS5 Monoclonal specifically detects RGS5 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen RGS5
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1C1
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias MST092, MST106, MST129, MSTP032, MSTP092, MSTP106, MSTP129, regulator of G-protein signaling 5, regulator of G-protein signalling 5
Gene Symbols RGS5
Host Species Mouse
Immunogen Full length recombinant protein of human RGS5 (NP_003608) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8490.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.