Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RHAMM/CD168 Rabbit anti-Human, Mouse, Clone: 3L10H9, Novus Biologicals™
SDP

Catalog No. NB165944 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165944 100 μg
NB165945 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB165944 Supplier Novus Biologicals Supplier No. NBP315424100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

RHAMM/CD168 Monoclonal antibody specifically detects RHAMM/CD168 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown

Specifications

Antigen RHAMM/CD168
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown
Classification Monoclonal
Clone 3L10H9
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin, Knockout Validated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias CD168, CD168 antigen, hyaluronan mediated motility receptor, hyaluronan-mediated motility receptor (RHAMM), IHABP, Intracellular hyaluronic acid-binding protein, MGC119494, Receptor for hyaluronan-mediated motility, RHAMMMGC119495
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human RHAMM/CD168 (O75330).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, CD Markers, Cell Biology, Immunology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 3161
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.