Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ RHC 80267
GREENER_CHOICE

Catalog No. 184210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-421-0 10 mg
18-425-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-421-0 Supplier Tocris Bioscience™ Supplier No. 1842/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Diacylglycerol lipase inhibitor

Inhibitor of diacylglycerol lipase (IC50 values are 1.1 and 4 μM in rat cardiac myocytes and canine platelets respectively). Weakly inhibits phospholipases C and A2. Potentiates acetylcholine evoked relaxation in mesenteric arteries by the inhibition of cholinesterase activity (IC50 = 4 μM).

Chemical Identifiers

CAS 83654-05-1
Molecular Formula C20H34N4O4

Specifications

Quantity 10 mg
Formula Weight 394.51
Purity >95%
Molecular Weight (g/mol) 394.52
Chemical Name or Material RHC 80267
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.