Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RHCE Antibody (BRIC69), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. N106887A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N106887A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N106887A30 Supplier Novus Biologicals Supplier No. NB10065887AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RHCE Monoclonal specifically detects RHCE in Human samples. It is validated for Flow Cytometry, Immunoprecipitation.

Specifications

Antigen RHCE
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone BRIC69
Conjugate Alexa Fluor 350
Dilution Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias blood group Rh(CE) polypeptide, blood group RhCcEe antigen, CD240CE, CD240CE antigen, MGC103977, RH, Rh blood group antigen Evans, Rh blood group C antigen, Rh blood group, CcEe antigens, Rh polypeptide 1, Rh polypeptide I, Rh30A, Rh4, RHC, RHCE blood group variant Crawford antigen Rh43, RHE, Rhesus blood group CE protein, Rhesus blood group E antigen, Rhesus blood group Rhce antigen, Rhesus blood group, CcEe antigens, Rhesus C/E antigens, Rhesus system C and E polypeptides, RhIVb(J), RHIXB, RhPI, RhVI, RhVIII, silenced Rh blood group CcEe antigen
Gene Symbols RHCE
Host Species Mouse
Immunogen Human erythrocytes
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 6006.0
Test Specificity Recognizes human erythrocytes of normal Rh types but does not recognize Rh null erythrocytes. NB100-65887 reacts with 31kD and 35-52kD proteins which were designated CD240DCE at the 7th leucocyte typing workshop.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.