Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RHCE Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB318215 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB318215 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB318215 Supplier Novus Biologicals Supplier No. NBP298927PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RHCE Polyclonal antibody specifically detects RHCE in Human samples. It is validated for Western Blot

Specifications

Antigen RHCE
Applications Western Blot
Classification Polyclonal
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias blood group Rh(CE) polypeptide, blood group RhCcEe antigen, CD240CE, CD240CE antigen, MGC103977, RH, Rh blood group antigen Evans, Rh blood group C antigen, Rh blood group, CcEe antigens, Rh polypeptide 1, Rh polypeptide I, Rh30A, Rh4, RHC, RHCE blood group variant Crawford antigen Rh43, RHE, Rhesus blood group CE protein, Rhesus blood group E antigen, Rhesus blood group Rhce antigen, Rhesus blood group, CcEe antigens, Rhesus C/E antigens, Rhesus system C and E polypeptides, RhIVb(J), RHIXB, RhPI, RhVI, RhVIII, silenced Rh blood group CcEe antigen
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human RHCE.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 6006
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.