Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RHCG Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB045733 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB045733 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB045733 Supplier Novus Biologicals Supplier No. NBP297617V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RHCG Polyclonal specifically detects RHCG in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen RHCG
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ammonium transporter Rh type C, CDRC2, chromosome 15 open reading frame 6, PDRC2C15orf6, Rh family type C glycoprotein, Rh family, C glycoprotein, Rh glycoprotein kidney, Rh type C glycoprotein, Rhesus blood group family type C glycoprotein, Rhesus blood group, C glycoprotein, RHGKTumor-related protein DRC2, SLC42A3
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human RHCG fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51458
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.