Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rheb Antibody (Rheb 5D5) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB395489 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB395489 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB395489 Supplier Novus Biologicals Supplier No. NBP2502730.025ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rheb Monoclonal antibody specifically detects Rheb in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen Rheb
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone Rheb 5D5
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Immunoprecipitation 5 μg/mL
Formulation PBS
Gene Alias GTP-binding protein Rheb, MGC111559, Ras homolog enriched in brainRHEB2Ras homolog enriched in brain 2
Host Species Mouse
Immunogen Human Rheb GST fusion
Purification Method Protein A purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, mTOR Pathway, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6009
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.