Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rheb Antibody (Rheb 5D5) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | Rheb |
---|---|
Clone | Rheb 5D5 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1 μg/mL, Immunoprecipitation 5 μg/mL |
Applications | Western Blot, Immunoprecipitation |
Description
Rheb Monoclonal antibody specifically detects Rheb in Human, Mouse samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
Rheb | |
1 mg/mL | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Cancer, mTOR Pathway, Signal Transduction | |
PBS | |
6009 | |
IgG2a | |
Protein A purified |
Rheb 5D5 | |
Western Blot 1 μg/mL, Immunoprecipitation 5 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
GTP-binding protein Rheb, MGC111559, Ras homolog enriched in brainRHEB2Ras homolog enriched in brain 2 | |
Human Rheb GST fusion | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title