Learn More
Thermo Scientific™ Rheodyne™ Replacement Rotor Seals for Injectors
Maintain performance of your Thermo Scientific™ Rheodyne™ Sample Injectors for HPLC with these replacement rotor seals.
$289.00 - $505.50
Specifications
Type Replacement Rotor Seals
View More Specs
Products 16
Catalog Number Mfr. No. For Use With (Equipment) Material Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Material Price Quantity & Availability  
03-052-241
View Documents
Thermo Scientific™
7010039
Rheodyne Injectors 7000/7010/7040/7067 Vespel
Each for $289.00
Only null left
Add to Cart
03-052-261
View Documents
Thermo Scientific™
7125047
Rheodyne Injectors 7125/7126 Vespel
Each for $505.50
Only null left
Add to Cart
03-050-413
View Documents
Thermo Scientific™
3725018
Rheodyne Injectors 3725/3725i/3725-038/3725i-038 PEEK N/A N/A N/A
03-052-244
View Documents
Thermo Scientific™
7010071
Rheodyne Injectors 7000/7010/7040 ETFE N/A N/A N/A
03-052-256
View Documents
Thermo Scientific™
7030003
Rheodyne Injector 7030 Vespel N/A N/A N/A
03-052-257
View Documents
Thermo Scientific™
7030015
Rheodyne Injector 7030 ETFE N/A N/A N/A
03-052-259
View Documents
Thermo Scientific™
7060070
Rheodyne Injectors 7060/7066 Vespel N/A N/A N/A
03-052-260
View Documents
Thermo Scientific™
7060074
Rheodyne Injectors 7060/7066/9060 ETFE N/A N/A N/A
03-052-264
View Documents
Thermo Scientific™
7125079
Rheodyne Injectors 7125/7126 ETFE N/A N/A N/A
03-052-269
View Documents
Thermo Scientific™
7410038
Rheodyne Injector 7410 Vespel N/A N/A N/A
03-052-271
View Documents
Thermo Scientific™
7410075
Rheodyne Injector 7410 ETFE N/A N/A N/A
03-052-273
View Documents
Thermo Scientific™
7413013
Rheodyne Injector 7413 Vespel N/A N/A N/A
03-052-365
View Documents
Thermo Scientific™
8125038
Rheodyne Injectors 8125/8126 Vespel N/A N/A N/A
03-052-369
View Documents
Thermo Scientific™
8125097
Rheodyne Injector 8125 ETFE N/A N/A N/A
03-052-394
View Documents
Thermo Scientific™
9010051
Rheodyne Injector 9010 ETFE N/A N/A N/A
03-052-389
View Documents
Thermo Scientific™
9125082
Rheodyne Injector 9125 ETFE N/A N/A N/A
Description
Description
Use Thermo Scientific™ Rheodyne™ Replacement Rotor Seals for Injectors to maintain the performance of your Rheodyne Sample Injectors for high-performance liquid chromatography (HPLC). These seals are suitable for popular Rheodyne injector models and are available in Vespel, PEEK or ETFE.
Specifications
Specifications
Replacement Rotor Seals
Videos
SDS
Documents
Documents
Product Certifications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.