Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RhoA Antibody, Novus Biologicals™
SDP

Catalog No. NB017894 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017894 0.025 mL
NB10091273 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017894 Supplier Novus Biologicals Supplier No. NB100912730.025ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

RhoA Polyclonal specifically detects RhoA in Human, Mouse, Primate, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen RhoA
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:300-1:3000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:500, Immunohistochemistry-Paraffin 1:300-1:3000
Formulation Whole antisera with No Preservative
Gene Accession No. P61586
Gene Alias h12, oncogene RHO H12, ras homolog gene family, member A, Rho cDNA clone 12, Rho12, RhoA, RHOH12ARH12ARHAAplysia ras-related homolog 12, small GTP binding protein RhoA, transforming protein RhoA
Gene Symbols RHOA
Host Species Rabbit
Immunogen A synthetic peptide as a part of human, rat, mouse, chicken, dog, bovine, frog, zebrafish, orangutan Transforming protein RhoA (Arha, Arha2) conjugated to an immunogenic carrier protein was used as the immunogen.
Molecular Weight of Antigen 22 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 387
Test Specificity RhOA.
Reconstitution Reconstitute with 0.1 ml sterilized water to desired concentration.
Target Species Human, Mouse, Mouse, Primate, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.