Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RhoA Rabbit anti-Human, Clone: 36, Novus Biologicals™
SDP

Catalog No. NB021674 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB021674 50 μg
NB021673 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB021674 Supplier Novus Biologicals Supplier No. NBP29010550UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

RhoA Monoclonal specifically detects RhoA in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen RhoA
Applications Flow Cytometry
Classification Monoclonal
Clone 36
Conjugate Unconjugated
Dilution Flow Cytometry 1:25-1:100
Formulation 0.2 um filtered solution in PBS
Gene Alias h12, oncogene RHO H12, ras homolog gene family, member A, Rho cDNA clone 12, Rho12, RhoA, RHOH12ARH12ARHAAplysia ras-related homolog 12, small GTP binding protein RhoA, transforming protein RhoA
Host Species Rabbit
Immunogen Recombinant Human RhoA Protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cancer, mTOR Pathway, Phospho Specific, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 387
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.