Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RhoB Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP103484
Description
RhoB Polyclonal specifically detects RhoB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
RhoB | |
Polyclonal | |
Unconjugated | |
Lyophilized from PBS with No Preservative | |
ARH6MSTP081, ARHBAplysia RAS-related homolog 6, h6, MST081, oncogene RHO H6, ras homolog gene family, member B, Rho cDNA clone 6, RHOH6RhoB, rho-related GTP-binding protein RhoB | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
Primary | |
RhOB. | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 10-50 ug/ml, Immunohistochemistry 10-50 ug/ml | |
P62745 | |
RHOB | |
A synthetic peptide as a part of human, rat, mouse RhoB conjugated to an immunogenic carrier protein was used as the immunogen. | |
0.5 mg | |
Angiogenesis | |
388 | |
Reconstitute with 0.5 ml sterilized water. Add glycerol (1:1) for additional stability. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction