Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RhoB Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Specifications
Antigen | RhoB |
---|---|
Concentration | LYOPH |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP103484
![]() |
Novus Biologicals
NBP103484 |
0.5 mg | N/A | N/A | N/A | ||||
Description
RhoB Polyclonal specifically detects RhoB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
RhoB | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Angiogenesis | |
P62745 | |
388 | |
IgG | |
Reconstitute with 0.5 ml sterilized water. Add glycerol (1:1) for additional stability. | |
RhOB. |
LYOPH | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
ARH6MSTP081, ARHBAplysia RAS-related homolog 6, h6, MST081, oncogene RHO H6, ras homolog gene family, member B, Rho cDNA clone 6, RHOH6RhoB, rho-related GTP-binding protein RhoB | |
RHOB | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title