Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RHOBTB2 Antibody (DBC2/3362), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB385259 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB385259 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB385259 Supplier Novus Biologicals Supplier No. NBP308659C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RHOBTB2 Monoclonal antibody specifically detects RHOBTB2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen RHOBTB2
Applications Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone DBC2/3362
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias DBC2KIAA0717Deleted in breast cancer 2 gene protein, p83, Rho-related BTB domain containing 2, rho-related BTB domain-containing protein 2
Host Species Mouse
Immunogen Human recombinant RHOBTB2 protein fragment (around aa554-604) (exact sequence is proprietary) (Uniprot: Q9BYZ6)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23221
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2c λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.