Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RhoC Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | RhoC |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
RhoC Polyclonal specifically detects RhoC in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RhoC | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
ARH9ARHCRho cDNA clone 9, H9, MGC1448, MGC61427, oncogene RHO H9, ras homolog gene family, member C, RAS-related homolog 9, RhoC, rhoC GTPase, RHOH9, rho-related GTP-binding protein RhoC, small GTP binding protein RhoC | |
RHOC | |
IgG | |
Affinity Purified |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
389 | |
This antibody was developed against a recombinant protein corresponding to amino acids: KDLRKAGTWRTGSVPLATLSAQPRPRRECGRCLRWPLGLASRSARTSVGGAVPFSEIPKAFPTC | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title