Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RhoC Antibody (OTI3A5), FITC, Novus Biologicals™
SDP

Catalog No. NBP273888F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273888F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273888F Supplier Novus Biologicals Supplier No. NBP273888F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RhoC Monoclonal specifically detects RhoC in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen RhoC
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI3A5
Conjugate FITC
Dilution Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ARH9ARHCRho cDNA clone 9, H9, MGC1448, MGC61427, oncogene RHO H9, ras homolog gene family, member C, RAS-related homolog 9, RhoC, rhoC GTPase, RHOH9, rho-related GTP-binding protein RhoC, small GTP binding protein RhoC
Gene Symbols RHOC
Host Species Mouse
Immunogen Synthetic peptide: VFEMATRAGLQVRKNKRRRG, corresponding to C terminal of human RhoC.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 389.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.