Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RhoC Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB092510 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB092510 20 μL
NB092509 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB092510 Supplier Novus Biologicals Supplier No. NBP30332220UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RhoC Polyclonal specifically detects RhoC in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.

Specifications

Antigen RhoC
Applications Western Blot, Immunohistochemistry, Immunofluorescence, KnockDown
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias ARH9ARHCRho cDNA clone 9, H9, MGC1448, MGC61427, oncogene RHO H9, ras homolog gene family, member C, RAS-related homolog 9, RhoC, rhoC GTPase, RHOH9, rho-related GTP-binding protein RhoC, small GTP binding protein RhoC
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-193 of human RhoC (NP_001036144.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 389
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.