Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RhoD Antibody (OTI2F7), PerCP, Novus Biologicals™
SDP

Catalog No. NB273889PCP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB273889PCP 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB273889PCP Supplier Novus Biologicals Supplier No. NBP273889PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RhoD Monoclonal specifically detects RhoD in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen RhoD
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2F7
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ARHDRHOM, ras homolog D, ras homolog gene family, member A, ras homolog gene family, member D, Rho, RhoD, RhoHP1, rho-related GTP-binding protein RhoD, Rho-related protein HP1
Gene Symbols RHOD
Host Species Mouse
Immunogen Full length recombinant protein of human RhoD (NP_055393) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline mTOR Pathway, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 29984.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.