Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodamine Antibody (11H10), Novus Biologicals™
SDP

Catalog No. NBP169961 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP169961 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP169961 Supplier Novus Biologicals Supplier No. NBP169961
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rhodamine Monoclonal specifically detects Rhodamine in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen Rhodamine
Applications Western Blot, ELISA, Immunohistochemistry, Dot Blot
Classification Monoclonal
Clone 11H10
Concentration 1.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, ELISA 1:10000-1:30000, Immunohistochemistry 1:1000-1:10000, Dot Blot, N/A
Gene Alias Rhodamine 6G, Rhodamine B
Host Species Mouse
Immunogen Rhodamine Antibody (11H10) was produced after repeated immunizations of balb/c mice with rhodamine conjugated KLH.
Purification Method Protein A purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity RHODAMINE Monoclonal Antibody was protein A purified and reacts specifically with Rhodamine and its derivatives. Rhodamine isomer 5 and isomer 6 are reactive as TAMRA, as well as TRITC conjugated proteins. No reaction is observed against Texas Red.
Target Species Non-species specific
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.