Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodium, 5% on alumina powder, C301099-5

Catalog No. p-7052540
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 7440-16-6 | Rh | 102.91 g/mol

Rhodium is a useful reagent for the conversation of nitriles to primary amines and the hydrogenation of aromatic rings, pyridines, aldehydes and sugars. It is also used for the reduction of oximes to amines and for the reduction of nitrobenzene to phenylhydroxylamine using hydrazine hydrate as the hydrogen source.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Rhodium is a useful reagent for the conversation of nitriles to primary amines and the hydrogenation of aromatic rings, pyridines, aldehydes and sugars. It is also used for the reduction of oximes to amines and for the reduction of nitrobenzene to phenylhydroxylamine using hydrazine hydrate as the hydrogen source.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with organic materials and flammable substances.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-16-6
Molecular Formula Rh
Molecular Weight (g/mol) 102.91
MDL Number MFCD00011201 MFCD03458392
InChI Key MHOVAHRLVXNVSD-UHFFFAOYSA-N
Synonym fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
PubChem CID 23948
ChEBI CHEBI:33359
IUPAC Name rhodium
SMILES [Rh]

Specifications

Color Gray
Physical Form Powder
Quantity 5 g
Solubility Information Insoluble in water.
Odor Odorless
Chemical Name or Material Rhodium, 5% on alumina powder, C301099-5
EINECSNumber 231-125-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.