Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rhodium(III) chloride hydrate, 99.95%, (trace metal basis)
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
Specifications
Specifications
| Melting Point | 100.0°C |
| Color | White |
| Physical Form | Crystalline Powder |
| Quantity | 250 mg |
| Assay Percent Range | 37.0 to 43% (Rh) |
| Linear Formula | RHCl3·xH2O |
| Solubility Information | Solubility in water: soluble. Other solubilities: soluble in alchol and hcl, insoluble in ether |
| Formula Weight | 209.26 |
| Percent Purity | 99.95% |
| Grade | Trace Metal Basis |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes serious eye damage. Harmful if swallowed. Suspected of causing genetic defects. May be corrosive to metals. Very toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Immediately |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction