Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodopsin Antibody (Rho 1D4), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB015093 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB015093 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB015093 Supplier Novus Biologicals Supplier No. NBP281059AF350
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Rhodopsin Monoclonal specifically detects Rhodopsin in Human, Mouse, Rat, Amphibian, Bovine, Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Rhodopsin
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone Rho 1D4
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CSNBAD1, MGC138309, OPN2MGC138311, opsin 2, rod pigment, opsin-2, retinitis pigmentosa 4, autosomal dominant, rhodopsin, RP4
Gene Symbols RHO
Host Species Rabbit
Immunogen Bleached bovine rod outer segment (ROS).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Phospho Specific, Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 6010
Test Specificity The antibody binds to the C-terminus of rhodopsin on the cytoplasmic side of disk membranes. The epitope has the following amino acid sequence: 'TETSQVAPA'. A limitation of the 1D4 tag, however, is that it has to be placed at the C-terminus of a protein. This is because the Rho1D4 monoclonal antibody requires a free carboxylate group for high affinity binding. Amidation of the carboxyl group lowers its immunoreactivity to the Rho1D4 antibody by over 100 fold.
Target Species Human, Mouse, Rat, Amphibian, Bovine, Zebrafish
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.