Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodopsin Antibody (Rho 1D4), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB342923 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342923 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342923 Supplier Novus Biologicals Supplier No. NBP281059MFV610
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Rhodopsin Monoclonal antibody specifically detects Rhodopsin in Human, Mouse, Rat, Amphibian, Bovine, Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Radioimmune Assays

Specifications

Antigen Rhodopsin
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone Rho 1D4
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Radioimmunoassay
Formulation 50mM Sodium Borate
Gene Alias CSNBAD1, MGC138309, OPN2MGC138311, opsin 2, rod pigment, opsin-2, retinitis pigmentosa 4, autosomal dominant, rhodopsin, RP4
Host Species Rabbit
Immunogen Bleached bovine rod outer segment (ROS).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Phospho Specific, Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 6010
Target Species Human, Mouse, Rat, Amphibian, Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.