Learn More
Rhodopsin, Rabbit anti-Human,Mouse,Rat,Amphibian,Bovine,Zebrafish, Clone: Rho 1D4, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Rhodopsin |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Radioimmune Assays (RIA) |
| Classification | Monoclonal |
| Clone | Rho 1D4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Radioimmunoassay |
| Formulation | PBS |
| Gene Alias | CSNBAD1, MGC138309, OPN2MGC138311, opsin 2, rod pigment, opsin-2, retinitis pigmentosa 4, autosomal dominant, rhodopsin, RP4 |
| Gene Symbols | RHO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.