Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodopsin Rabbit anti-Mouse, Rat, Clone: 5N3G9, Novus Biologicals™
SDP

Catalog No. NB168402 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168402 20 μg
NB168401 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB168402 Supplier Novus Biologicals Supplier No. NBP31665520UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Rhodopsin Monoclonal antibody specifically detects Rhodopsin in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Rhodopsin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5N3G9
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias CSNBAD1, MGC138309, OPN2MGC138311, opsin 2, rod pigment, opsin-2, retinitis pigmentosa 4, autosomal dominant, rhodopsin, RP4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 249-348 of human Rhodopsin (P08100).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline GPCR, Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 6010
Target Species Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.