Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RhoF Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB054596 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB054596 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB054596 Supplier Novus Biologicals Supplier No. NBP298649R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RhoF Polyclonal specifically detects RhoF in Human samples. It is validated for Western Blot.

Specifications

Antigen RhoF
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias FLJ20247, ras homolog gene family, member F (in filopodia), Rho family GTPase Rif, Rho in filopodia, rho-related GTP-binding protein RhoF, RIFARHF
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human RhoF fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 54509
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.