Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RhoG Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB342298 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342298 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342298 Supplier Novus Biologicals Supplier No. NBP281812MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RhoG Polyclonal antibody specifically detects RhoG in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen RhoG
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ARHGMGC125835, MGC125836, ras homolog gene family, member G (rho G), RhoG, rho-related GTP-binding protein RhoG
Host Species Rabbit
Immunogen RHOG antibody was raised against a 15 amino acid peptide near the amino terminus of human RHOG. The immunogen is located within amino acids 40 - 90 of RHOG. Amino Acid Squence: WDTAGQEEYDRLRTL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 391
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.