Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RhoGDI Antibody (OTI1F2) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP273890 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273890 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273890 Supplier Novus Biologicals Supplier No. NBP273890
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RhoGDI Monoclonal specifically detects RhoGDI in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.

Specifications

Antigen RhoGDI
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone OTI1F2
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:200-1000, Flow Cytometry 1:100, CyTOF-ready
Formulation Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Gene Alias GDIA1, MGC117248, Rho GDI 1, Rho GDP dissociation inhibitor (GDI) alpha, rho GDP-dissociation inhibitor 1, RHOGDI, Rho-GDI alpha, RHOGDI-1
Gene Symbols ARHGDIA
Host Species Mouse
Immunogen Full length human recombinant protein of human ARHGDIA (NP_004300) produced in HEK293T cell.
Molecular Weight of Antigen 23 kDa
Purification Method Affinity Purified
Quantity 100 μg
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 396
Reconstitution Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Target Species Human, Mouse, Rat, Canine, Primate, Monkey
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.