Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RhoJ Antibody (OTI1D7), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP273892IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273892IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273892IR Supplier Novus Biologicals Supplier No. NBP273892IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RhoJ Monoclonal specifically detects RhoJ in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen RhoJ
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1D7
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ARHJTC10B, FLJ14445, ras homolog gene family, member J, RASL7B, Ras-like protein family member 7B, RAS-like, family 7, member B, RHOI, rho-related GTP-binding protein RhoJ, Tc10-like GTP-binding protein, TC10-like Rho GTPase, TCLMGC34777
Gene Symbols RHOJ
Host Species Mouse
Immunogen Full length recombinant protein of human RhoJ (NP_065714) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 57381.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.