Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhox10 Antibody, Novus Biologicals™
SDP

Catalog No. NBP223834 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223834 0.1 mL
NBP223834SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP223834 Supplier Novus Biologicals Supplier No. NBP223834
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rhox10 Polyclonal specifically detects Rhox10 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Rhox10
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1: 1000-1:2000
Formulation 0.1 ml crude whole Rabbit sera with 50% Glycerol with 0.025% Sodium Azide
Gene Alias reproductive homeobox 10, Rhox10 reproductive homeobox 10
Host Species Rabbit
Immunogen Within the range of amino acids 90-118 of mouse Rhox-10 protein were used as the immunogen.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 434769
Test Specificity The amino acid sequence used as immunogen is 100% homologous in mouse and rat and 82% homologous in human, rabbit and dog.
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.