Learn More
Description
Specifications
Specifications
| Antigen | Rhox11 |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1: 1000-1:2000, Immunohistochemistry-Paraffin 1: 100 |
| Formulation | 0.1 ml crude whole Rabbit sera with 50% Glycerol with 0.025% Sodium Azide |
| Gene Alias | BC049711, Gm39,11, MGC58663, reproductive homeobox 11 |
| Gene Symbols | RHOX11 |
| Host Species | Rabbit |
| Immunogen | Within the range of amino acids 139-171 of mouse Rhox-11 protein were used as the immunogen. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
