Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RHOXF1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP23379025UL
Description
RHOXF1 Polyclonal specifically detects RHOXF1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RHOXF1 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Q8NHV9 | |
RHOXF1 | |
This antibody was developed against a recombinant protein corresponding to amino acids: AMEGPQPENMQPRTRRTKFTLLQVEELESVFRHTQYPDVPTRRELAENLGVTEDKVRVWF | |
25 μL | |
Signal Transduction | |
158800 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
MGC119030, OTEXMGC119033, Ovary-, testis- and epididymis-expressed gene protein, Paired-like homeobox protein PEPP-1, PEPP subfamily gene 1, PEPP1paired-like homeobox protein OTEX, rhox homeobox family member 1, Rhox homeobox family, member 1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction