Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RHOXF1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | RHOXF1 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
RHOXF1 Polyclonal specifically detects RHOXF1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RHOXF1 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
Q8NHV9 | |
158800 | |
This antibody was developed against a recombinant protein corresponding to amino acids: AMEGPQPENMQPRTRRTKFTLLQVEELESVFRHTQYPDVPTRRELAENLGVTEDKVRVWF | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
MGC119030, OTEXMGC119033, Ovary-, testis- and epididymis-expressed gene protein, Paired-like homeobox protein PEPP-1, PEPP subfamily gene 1, PEPP1paired-like homeobox protein OTEX, rhox homeobox family member 1, Rhox homeobox family, member 1 | |
RHOXF1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title