Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RHOXF2 Antibody (PCRP-RHOXF2-1D7) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB119255 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119255 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119255 Supplier Novus Biologicals Supplier No. NBP308529
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RHOXF2 Monoclonal specifically detects RHOXF2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.

Specifications

Antigen RHOXF2
Applications Flow Cytometry, Immunofluorescence, Peptide Array
Classification Monoclonal
Clone PCRP-RHOXF2-1D7
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry : 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence : 1-2 ug/mL, Protein Array
Formulation 10mM PBS
Gene Alias CT107, Paired-like homeobox protein PEPP-2, PEPP subfamily gene 2, PEPP-2, PEPP2cancer/testis antigen 107, rhox homeobox family member 2, Rhox homeobox family, member 2, Testis homeobox gene 1, THG1homeobox protein from AL590526
Host Species Mouse
Immunogen Recombinant full-length human RHOXF2 protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84528
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.