Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RHOXF2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB125815 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB125815 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB125815 Supplier Novus Biologicals Supplier No. NBP310457100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RHOXF2 Polyclonal specifically detects RHOXF2 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen RHOXF2
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry
Formulation PBS buffer, 2% sucrose
Gene Alias CT107, Paired-like homeobox protein PEPP-2, PEPP subfamily gene 2, PEPP-2, PEPP2cancer/testis antigen 107, rhox homeobox family member 2, Rhox homeobox family, member 2, Testis homeobox gene 1, THG1homeobox protein from AL590526
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the middle region of human RHOXF2 (NP_115887). Peptide sequence DQSEKEPGQQYSRPQGAVGGLEPGNAQQPNVHAFTPLQLQELERIFQREQ
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84528
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.