Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ RIA Vial
SDPSCI_ED

Catalog No. S15682 Shop All United Scientific Supplies, Inc Products
Click to view available options
Material:
Polypropylene
Polystyrene

Catalog No. S15682

Supplier: United Scientific™ P10301

Only null left
Add to Cart
Add to Cart

Caps and tubes sold separately and are used in radionimmunoassay.

  • RIA vials are generally used in radioimmunoassay, a scientific method used to test antigens without the need to use a bioassay.
  • The polypropylene vials are translucent and can withstand 3000 RCF during centrifugation.
  • Can be used with common acids, alkalies, and solvents at room temperature. Autoclavable.
  • The polystyrene vials are transparent and can withstand 1400 RCF during centrifugation.
  • Can be used with mild bases and weak acids, but not organic solvents.
  • Non-autoclavable.
  • Caps are sold separately and are compatible with both versions of vials.

Specifications

Color Clear
Diameter (Metric) Outer 12 mm
Material Polypropylene
Pre-slit No
Sterility Non-sterile
Volume (Metric) 5 mL
Autoclavable Autoclavable
Closure Type Screw Cap (Not Included)
Height (Metric) 75 mm
Quantity 500/Pk.
Type Vial
Preservative No
Marking Spot No
Vial Type RIA
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.